siRNA Details
Gene Symbol: Ccdc172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014169.2 | NP_001014191.1 | 3 | 192 | ||
XM_006231674.2 | XP_006231736.1 | 3 | 208 | |||
XM_008760539.1 | XP_008758761.1 | 2 | 481 | |||
XM_008760540.1 | XP_008758762.1 | 2 | 481 | |||
XM_008760541.1 | XP_008758763.1 | 2 | 481 | |||
XM_008760542.1 | XP_008758764.1 | 2 | 481 | |||
GenBank mRNA | BC079324.1 | - | 2 | 409 | 7135457 | |
CV104909.1 | - | 3 | 192 |
