siRNA Details
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108535.1 | NP_001102005.1 | 3 | 364 | ||
XM_006256462.1 | XP_006256524.1 | 3 | 310 | |||
XM_006256463.1 | XP_006256525.1 | 3 | 314 | |||
XM_006256464.2 | XP_006256526.1 | 3 | 311 | |||
XM_008772893.1 | XP_008771115.1 | 1 | 1329 | |||
XM_008772894.1 | XP_008771116.1 | 3 | 308 | |||
XM_008772895.1 | XP_008771117.1 | 1 | 1328 | |||
XM_008772896.1 | XP_008771118.1 | 1 | 1329 |
Gene Symbol: LOC100911233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751966.3 | XP_003752014.1 | 3 | 313 |
