siRNA Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108539.3 | NP_001102009.2 | 24 | 2650 | ||
XM_006256595.2 | XP_006256657.1 | 24 | 2924 | |||
XM_006256597.2 | XP_006256659.1 | 23 | 2792 | |||
XM_006256598.1 | XP_006256660.1 | 19 | 2262 | |||
GenBank mRNA | CB327407.1 | - | 1 | 693 | ||
CO402715.1 | - | 6 | 562 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172473, AK042879, AK029162, BC120695, BC025227, and others.
Gene Ontology Categories:
