siRNA Details
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012177.1 | NP_001012177.1 | 6 | 710 | ||
XM_008760678.1 | XP_008758900.1 | 6 | 896 | |||
GenBank mRNA | BC083581.1 | - | 6 | 710 | 7129446 |

More Information
Other Species Targeted

Gene Ontology Categories:
