siRNA Details
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047910.1 | NP_001041375.1 | 10, 11 | 1389 | ||
GenBank mRNA | AY325200.1 | - | 10, 11 | 1389 |

More Information
Other Species Targeted

Gene Ontology Categories:
