siRNA Details
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047910.1 | NP_001041375.1 | 5 | 470 | ||
GenBank mRNA | AY325200.1 | - | 5 | 470 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028140, AK077957, BC113183, BC113790, AK032080, and others.
Gene Ontology Categories:
