siRNA Details
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115029.2 | NP_001108501.2 | 3, 4 | 719 | ||
XM_006232473.1 | XP_006232535.1 | 3, 4 | 381 | |||
XM_006232474.2 | XP_006232536.1 | 2, 3 | 593 | |||
XM_006232475.2 | XP_006232537.1 | 2, 3 | 244 | |||
GenBank mRNA | BC107462.1 | - | 3, 4 | 683 | ||
BC158627.1 | - | 3, 4 | 719 | 7455378 |
