siRNA Details
Gene Symbol: Fga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008724.1 | NP_001008724.1 | 4 | 288 | ||
NM_052797.1 | NP_434684.1 | 3 | 288 | |||
XM_006232532.2 | XP_006232594.1 | 3 | 332 | |||
XM_006232533.2 | XP_006232595.1 | 3 | 324 | |||
GenBank mRNA | AY310161.1 | - | 4 | 288 | ||
BC127465.1 | - | 3 | 288 | 8375711 | ||
FQ209852.1 | - | 3 | 306 |
