siRNA Details
Gene Symbol: S100a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108557.2 | NP_001102027.1 | 2 | 84 | ||
XM_006232717.2 | XP_006232779.1 | 2 | 158 | |||
XM_006232718.1 | XP_006232780.1 | 2 | 75 | |||
GenBank mRNA | BC158598.1 | - | 2 | 84 | 7127572 | |
FQ209774.1 | - | 2 | 86 | |||
FQ214291.1 | - | 2 | 67 | |||
FQ214430.1 | - | 2 | 80 | |||
FQ229313.1 | - | 2 | 92 |
