siRNA Details
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025731.1 | NP_001020902.1 | 7, 8 | 1186 | ||
XM_006233836.2 | XP_006233898.1 | 7, 8 | 1222 | |||
XM_006233838.2 | XP_006233900.1 | 7, 8 | 1221 | |||
GenBank mRNA | BC099088.1 | - | 7, 8 | 1186 | 7456179 | |
FQ217346.1 | - | 7, 8 | 1207 |
