siRNA Details
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108579.1 | NP_001102049.1 | 6 | 1266 | ||
XM_008761759.1 | XP_008759981.1 | 5 | 2133 | |||
XM_008761760.1 | XP_008759982.1 | 4 | 2142 | |||
GenBank mRNA | BC158641.1 | - | 5 | 775 |
