siRNA Details
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014180.1 | NP_001014202.1 | 8, 9 | 805 | ||
XM_006234585.2 | XP_006234647.1 | 8, 9 | 883 | |||
GenBank mRNA | BC083756.1 | - | 8, 9 | 805 | 7192721 |
