siRNA Details
Gene Symbol: Trpc4ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100748.1 | NP_001094218.1 | 7 | 764 | ||
GenBank mRNA | AY724540.1 | - | 7 | 736 | ||
BC087662.1 | - | 7 | 717 | |||
BC166758.1 | - | 7 | 737 | 7930596 |
