siRNA Details
Gene Symbol: Myt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108615.1 | NP_001102085.1 | 5 | 2030 | ||
XM_006235782.1 | XP_006235844.1 | 11 | 1847 | |||
XM_006235783.2 | XP_006235845.1 | 11 | 2081 | |||
XM_006235784.1 | XP_006235846.1 | 11 | 1847 | |||
XM_006235785.2 | XP_006235847.1 | 11 | 1798 | |||
XM_006235786.2 | XP_006235848.1 | 11 | 1847 | |||
XM_006235787.2 | XP_006235849.1 | 8 | 1668 | |||
XM_008762522.1 | XP_008760744.1 | 11 | 1847 |
