siRNA Details
Gene Symbol: Sqrdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047913.2 | NP_001041378.2 | 1 | 243 | ||
XM_006234887.2 | XP_006234949.1 | 2 | 315 | |||
GenBank mRNA | AY325246.1 | - | 2 | 77 | ||
BC107459.1 | - | 2 | 251 | |||
BC158559.1 | - | 1 | 243 | 7456444 |
