siRNA Details
Gene Symbol: Bend5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108672.1 | NP_001102142.1 | 4 | 1032 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026279, BC051512, AK156708, BC037633, AK075885, and others.
