siRNA Details
Gene Symbol: Sync | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108683.1 | NP_001102153.1 | 2 | 1029 | ||
XM_006238943.2 | XP_006239005.1 | 2 | 1099 | |||
XM_006238944.2 | XP_006239006.1 | 2 | 1098 | |||
XM_006238945.2 | XP_006239007.1 | 2 | 1099 |

More Information
Other Species Targeted

Gene Ontology Categories:
