siRNA Details
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108690.1 | NP_001102160.1 | 18 | 2110 | ||
XM_006239245.2 | XP_006239307.1 | 19 | 2141 | |||
GenBank mRNA | FQ216974.1 | - | 19 | 2100 | ||
FQ218240.1 | - | 3 | 265 |
