siRNA Details
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014194.1 | NP_001014216.1 | 2 | 178 | ||
GenBank mRNA | BC089875.1 | - | 2 | 178 | 7325475 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014194.1 | NP_001014216.1 | 2 | 178 | ||
GenBank mRNA | BC089875.1 | - | 2 | 178 | 7325475 |