siRNA Details
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108702.2 | NP_001102172.2 | 9 | 1199 | ||
XM_008764486.1 | XP_008762708.1 | 10 | 1575 | |||
XM_008764487.1 | XP_008762709.1 | 10 | 1572 | |||
XM_008764488.1 | XP_008762710.1 | 9 | 1480 | |||
GenBank mRNA | BC166846.1 | - | 9 | 1199 | 9030077 |
