siRNA Details
Gene Symbol: Btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108720.1 | NP_001102190.1 | 9 | 3001 | ||
XM_008764818.1 | XP_008763040.1 | 10 | 3486 | |||
XM_008764819.1 | XP_008763041.1 | 11 | 3628 |
