siRNA Details
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047915.1 | NP_001041380.1 | 7, 8 | 946 | ||
XM_008765004.1 | XP_008763226.1 | 17, 18 | 2014 | |||
GenBank mRNA | AY390379.1 | - | 7, 8 | 946 | ||
FQ211964.1 | - | 9, 10 | 1166 |
