siRNA Details
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040190.1 | NP_001035280.1 | 12, 13 | 1579 | ||
GenBank mRNA | AY611623.1 | - | 12, 13 | 1579 | ||
BC128695.1 | - | 12, 13 | 1431 | 8366437 |
