siRNA Details
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108736.2 | NP_001102206.2 | 5 | 929 | ||
XM_006240973.2 | XP_006241035.1 | 5 | 962 | |||
XM_006240974.2 | XP_006241036.1 | 5 | 995 | |||
XM_006240975.2 | XP_006241037.1 | 5 | 1916 | |||
GenBank mRNA | BC160828.1 | - | 5 | 929 | 7453186 | |
CK597826.1 | - | 194 | ||||
CK603499.1 | - | 1 | 463 |
