siRNA Details
Gene Symbol: Derl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014202.1 | NP_001014224.1 | 3, 4 | 376 | ||
GenBank mRNA | BC085877.1 | - | 3, 4 | 376 | 7191715 | |
FQ218639.1 | - | 3, 4 | 413 | |||
FQ220628.1 | - | 3, 4 | 415 |
