siRNA Details
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130563.1 | NP_001124035.1 | 16 | 2194 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130563.1 | NP_001124035.1 | 16 | 2194 |