siRNA Details
Gene Symbol: Adck5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135798.1 | NP_001129270.1 | 14 | 1711 | ||
XM_006241827.2 | XP_006241889.1 | 14 | 1615 | |||
GenBank mRNA | BC168693.1 | - | 14 | 1699 | 7935927 | |
BP468309.1 | - | 2 | 276 |
