siRNA Details
Gene Symbol: LOC102552166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766604.1 | XP_008764826.1 | 4 | 878 |
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134520.1 | NP_001127992.1 | 5 | 648 |

More Information
Other Species Targeted

Gene Ontology Categories:
