siRNA Details
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007754.1 | NP_001007755.1 | 4, 5 | 701 | ||
NM_001037555.1 | NP_001032644.1 | 5, 6 | 893 | |||
GenBank mRNA | AB202124.1 | - | 5, 6 | 893 | ||
BC079237.1 | - | 4, 5 | 701 | 7132629 |
