siRNA Details
Gene Symbol: Gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191885.1 | NP_001178814.1 | 5 | 808 | ||
XM_008766645.1 | XP_008764867.1 | 5 | 829 | |||
GenBank mRNA | FQ211779.1 | - | 768 | |||
FQ217318.1 | - | 708 |
