siRNA Details
Gene Symbol: Csrnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108786.1 | NP_001102256.1 | 5 | 1581 | ||
XM_006244119.2 | XP_006244181.1 | 5 | 2248 | |||
XM_006244120.2 | XP_006244182.1 | 5 | 1853 | |||
XM_006244121.1 | XP_006244183.1 | 5 | 4004 | |||
XM_006244122.2 | XP_006244184.1 | 5 | 1598 | |||
XM_008766688.1 | XP_008764910.1 | 4 | 4921 | |||
XM_008766689.1 | XP_008764911.1 | 5 | 3865 |
