siRNA Details
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013227.2 | NP_001013245.1 | 7 | 568 | ||
GenBank mRNA | BC063182.1 | - | 7 | 572 | 6887869 | |
FQ215552.1 | - | 7 | 567 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013227.2 | NP_001013245.1 | 7 | 568 | ||
GenBank mRNA | BC063182.1 | - | 7 | 572 | 6887869 | |
FQ215552.1 | - | 7 | 567 |