siRNA Details
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025747.1 | NP_001020918.1 | 2 | 165 | ||
XM_006257077.2 | XP_006257139.1 | 4 | 322 | |||
GenBank mRNA | BC097987.1 | - | 2 | 165 | 7444864 |
