siRNA Details
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014225.1 | NP_001014247.1 | 9 | 1207 | ||
XM_006245832.2 | XP_006245894.1 | 9 | 970 | |||
XM_008767512.1 | XP_008765734.1 | 9 | 903 | |||
XM_008767513.1 | XP_008765735.1 | 9 | 1265 | |||
XM_008767514.1 | XP_008765736.1 | 8 | 807 | |||
XM_008767515.1 | XP_008765737.1 | 8 | 811 | |||
XM_008767516.1 | XP_008765738.1 | 8 | 809 | |||
XM_008767517.1 | XP_008765739.1 | 7 | 672 | |||
XM_008767518.1 | XP_008765740.1 | 7 | 729 | |||
GenBank mRNA | BC079022.1 | - | 9 | 1207 | 7113868 |
