siRNA Details
Gene Symbol: LOC690286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001073964.3 | XP_001073964.2 | 2 | 881 | ||
XM_006247136.1 | XP_006247198.1 | 2 | 881 |
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S79820.1 | - | 2 | 416 |
