siRNA Details
Gene Symbol: Meox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108837.1 | NP_001102307.1 | 3 | 733 | ||
XM_008768090.1 | XP_008766312.1 | 2 | 993 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Meox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108837.1 | NP_001102307.1 | 3 | 733 | ||
XM_008768090.1 | XP_008766312.1 | 2 | 993 |