siRNA Details
Gene Symbol: Arl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108842.1 | NP_001102312.1 | 5 | 586 | ||
XM_006248191.2 | XP_006248253.1 | 5 | 861 | |||
XM_006248192.2 | XP_006248254.1 | 5 | 711 | |||
XM_006248193.2 | XP_006248255.1 | 5 | 860 | |||
GenBank mRNA | BC162029.1 | - | 5 | 707 | 9100375 |
