siRNA Details
Gene Symbol: Rnf144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108881.1 | NP_001102351.1 | 3, 4 | 449 | ||
XM_006253767.2 | XP_006253829.1 | 3, 4 | 882 | |||
XM_008771549.1 | XP_008769771.1 | 3, 4 | 887 | |||
XM_008771550.1 | XP_008769772.1 | 3, 4 | 975 |
