siRNA Details
Gene Symbol: Gypc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013233.1 | NP_001013251.1 | 3 | 401 | ||
XM_006254599.2 | XP_006254661.1 | 3 | 4371 | |||
GenBank mRNA | AY234182.1 | - | 3 | 401 | ||
BC103641.1 | - | 3 | 314 | 7189007 |
