siRNA Details
Gene Symbol: Reep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108888.2 | NP_001102358.2 | 5 | 1019 | ||
GenBank mRNA | BC167013.1 | - | 5 | 944 | 9032688 | |
FQ213789.1 | - | 5 | 952 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Reep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108888.2 | NP_001102358.2 | 5 | 1019 | ||
GenBank mRNA | BC167013.1 | - | 5 | 944 | 9032688 | |
FQ213789.1 | - | 5 | 952 |