siRNA Details
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013234.1 | NP_001013252.1 | 3 | 719 | ||
XM_008772414.1 | XP_008770636.1 | 3 | 249 | |||
XM_008772415.1 | XP_008770637.1 | 1 | 33 | |||
XM_008772416.1 | XP_008770638.1 | 1 | 29 | |||
GenBank mRNA | BC086345.1 | - | 3 | 249 | 7302028 | |
BC086539.1 | - | 3 | 719 | 7114943 | ||
FQ227287.1 | - | 1 | 78 |
