siRNA Details
Gene Symbol: Xrra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748930.3 | XP_003748978.1 | 8 | 901 | ||
XM_006229821.2 | XP_006229883.1 | 7 | 803 | |||
GenBank mRNA | BC158768.1 | - | 8 | 743 | 7115524 |
