siRNA Details
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013238.1 | NP_001013256.1 | 3, 4 | 339 | ||
XM_006256253.2 | XP_006256315.1 | 4, 5 | 529 | |||
GenBank mRNA | BC061813.1 | - | 3, 4 | 339 | 5598663 | |
FQ218172.1 | - | 3, 4 | 351 |
