siRNA Details
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012218.1 | NP_001012218.1 | 5 | 772 | ||
XM_006232181.2 | XP_006232243.1 | 5 | 810 | |||
XM_006232182.2 | XP_006232244.1 | 5 | 810 | |||
GenBank mRNA | BC087100.1 | - | 5 | 772 | 7192411 |
