siRNA Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015032.3 | NP_001015032.2 | 2 | 739 | ||
XM_006234290.2 | XP_006234352.1 | 3 | 974 | |||
GenBank mRNA | AB040674.1 | - | 2 | 628 | ||
BC101886.1 | - | 2 | 742 | 7929522 |
