siRNA Details
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005892.2 | NP_001005892.1 | 4 | 591 | ||
XM_006234532.2 | XP_006234594.1 | 4 | 1470 | |||
GenBank mRNA | AB154364.1 | - | 4 | 454 | ||
BC091185.1 | - | 4 | 591 | 7366435 |
