siRNA Details
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108964.1 | NP_001102434.1 | 5, 6 | 521 | ||
XM_006235698.2 | XP_006235760.1 | 6, 7 | 681 | |||
XM_006235701.2 | XP_006235763.1 | 6, 7 | 681 | |||
XM_008762524.1 | XP_008760746.1 | 6, 7 | 681 | |||
XM_008762525.1 | XP_008760747.1 | 6, 7 | 1369 | |||
XM_008762527.1 | XP_008760749.1 | 6, 7 | 915 |
