siRNA Details
Gene Symbol: Cdc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013240.2 | NP_001013258.1 | 4 | 441 | ||
NM_001267054.1 | NP_001253983.1 | 4 | 369 | |||
NM_001267055.1 | NP_001253984.1 | 3 | 333 | |||
XM_006238241.2 | XP_006238303.1 | 4 | 457 | |||
XM_006238242.2 | XP_006238304.1 | 3 | 345 | |||
XM_008763796.1 | XP_008762018.1 | 4 | 380 | |||
GenBank mRNA | AY156922.1 | - | 4 | 437 | ||
BC086343.1 | - | 4 | 430 | 7301739 | ||
DY317988.1 | - | 3 | 328 | |||
FM103539.1 | - | 3 | 269 | |||
FQ217142.1 | - | 3 | 327 |
