siRNA Details
Gene Symbol: Tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013244.1 | NP_001013262.1 | 1, 2 | 230 | ||
GenBank mRNA | BC089906.1 | - | 1, 2 | 230 | 7123552 | |
FQ215999.1 | - | 1, 2 | 245 | |||
FQ217050.1 | - | 1, 2 | 245 |
