siRNA Details
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037097.1 | NP_001032174.1 | 12 | 1751 | ||
XM_008764823.1 | XP_008763045.1 | 12 | 1849 | |||
GenBank mRNA | BC107662.1 | - | 12 | 1751 | 7936642 |
